Learn More
Labnet™ ProBlot™ Economy Rocker
Product Code. 11491967
Change view
Click to view available options
Platform Style:
Double Platform
Single Platform
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Platform Style unitSize
11491967 Double Platform Each
11466707 Single Platform Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11491967 Supplier Labnet™ Supplier No. S2025DB230V
Only null left
Add to basket
Add to basket
ProBlot™ Economy Rocker is a high capacity rocker designed for durability.
  • The Labnet Economy Rockers, ProBlot™ 25 and 25XL high capacity bench rockers are basic bench rocking platforms that have been designed for a long life and trouble-free service.
    • Staining, hybridization, washing, etc.
    • Compact size, low profile
    • Extremely quiet
    • Double platform option expands usable space
  • Bench rocker speed is adjustable to provide the gentle motion required for staining fragile gels as well as the more vigorous action necessary for washing blots. The rocking movement is fixed at ±7°.
  • The platform surfaces are covered with nonslip rubber pads to prevent objects from moving during operation. Elastic tie downs are available to secure in place bags, boxes, tubes and other containers. The platform assemblies pivot on permanently lubricated ball bearings for quiet, maintenance-free operation.
  • The ProBlot™ 25 platform measures 28.9 x 20.3 cm, while the ProBlot™ 25XL platform is slightly larger at 30 x 30 cm.
  • For increasing the work area without sacrificing valuable bench space, a second platform can be stacked on the first. The ProBlot™ 25 and 25XL Rockers can be used safely in temperature controlled environments up to 65°C. Their low profile and compact size make them perfect for use in small incubators.
Specifications
Tilt Angle
Voltage 230 V
Temperature Range (Metric) 4°C to 65°C
Frequency 50/60 Hz
Dimensions (L x W x H) 32 x 29 x 14.5 cm (12.6 x 11.4 x 5.7 in.)
Platform Style Double Platform
Timer Range 0 to 2 hr. or Continuous