missing translation for 'onlineSavingsMsg'
Learn More

Pro-xylane, MedChemExpress

Product Code. 16490517
Encompass_Preferred
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 16490517

Brand: Medchem Express HY108036

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pro-xylane (Hydroxypropyl tetrahydropyrantriol) is a biologically active C-glycoside in aqueous media, acts as an activator of glycosaminoglycans (GAGs) biosynthesis. Pro-xylane is the first example of 'Green' chemical used in cosmetic.

Pro-xylane (Hydroxypropyl tetrahydropyrantriol) is a biologically active C-glycoside in aqueous media, acts as an activator of glycosaminoglycans (GAGs) biosynthesis. Pro-xylane is the first example of 'Green' chemical used in cosmetic.

Specifications

Chemical Name or Material Pro-xylane
Physical Form Liquid
CAS 439685-79-7
Color Colorless
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C8H16O5
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym Hydroxypropyl tetrahydropyrantriol
Solubility Information H2O : 250 mg/mL (1300.66 mM; Need ultrasonic) ∣DMSO : ≥ 83.33 mg/mL (433.54 mM)
SMILES CC(O)C[C@H]1[C@@H]([C@H]([C@H](O)CO1)O)O
Molecular Weight (g/mol) 192.21
Formula Weight 192.21
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.