Learn More
Biochrom Printer Accessory for NanoVue Plus and GeneQuant 1300/100
Click to view available options
Type:
Internal Printer
Unit Size:
Each
Description
- Printer Accessory for GeneQuant 1300/100 and NanoVue Plus spectrophotometers
- Optional built-in printer or output data via USB to a PC for printing or storage
- Easy to install
- Suitable for determining the concentration (in μg/mL, ng/μL, μg/L, Pmol/μL, and Pmol) and Purity of Nucleic Acids After Hybridization, PCR, and Plasmid DNA Minipreps
Specifications
Specifications
| Connections | Wireless Bluetooth |
| Type | Internal Printer |
| For Use With (Application) | Nucleic Acid Purification, Protein Assays, CyDye, and Cell Culture Density Measurements |
| Includes | Optional Built-in Printer or Output Data Via USB to a PC for Printing or Storage |