missing translation for 'onlineSavingsMsg'
Learn More

Prinomastat, MedChemExpress

Product Code. 16417737
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16417737 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16417737 Supplier Medchem Express Supplier No. HY12170

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Prinomastat (AG3340) is a broad spectrum, potent, orally active metalloproteinase (MMP) inhibitor with IC50s of 79, 6.3 and 5.0 nM for MMP-1, MMP-3 and MMP-9, respectively. Prinomastat inhibits MMP-2, MMP-3 and MMP-9 with Kis of 0.05 nM, 0.3 nM and 0.26 nM, respectively. Prinomastat crosses blood-brain barrier. Antitumor avtivity.

Prinomastat (AG3340) is a broad spectrum, potent, orally active metalloproteinase (MMP) inhibitor with IC50s of 79, 6.3 and 5.0 nM for MMP-1, MMP-3 and MMP-9, respectively. Prinomastat inhibits MMP-2, MMP-3 and MMP-9 with Kis of 0.05 nM, 0.3 nM and 0.26 nM, respectively. Prinomastat crosses blood-brain barrier. Antitumor avtivity.

Specifications

Chemical Name or Material Prinomastat
CAS 192329-42-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H21N3O5S2
Quantity 5 mg
Synonym AG3340 KB-R9896
Solubility Information DMSO : < 1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble or slightly soluble)
SMILES O=C([C@@H]1N(S(=O)(C2=CC=C(OC3=CC=NC=C3)C=C2)=O)CCSC1(C)C)NO
Molecular Weight (g/mol) 423.51
Formula Weight 423.51
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.