missing translation for 'onlineSavingsMsg'
Learn More

PRICKLE3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16878864 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16878864 20 μL 20µL
16868864 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16878864 Supplier Proteintech Supplier No. 190981AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LIM domain only 6 is a three LIM domain-containing protein. The LIM domain is a cysteine-rich sequence motif that binds zinc atoms to form a specific protein-binding interface for protein-protein interactions.

Specifications

Antigen PRICKLE3
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PRICKLE3
Gene Accession No. O43900, Q80VL3
Gene Alias LIM domain only protein 6, LMO 6, LMO6, prickle homolog 3 (Drosophila), Prickle like protein 3, PRICKLE3, Triple LIM domain protein 6
Gene Symbols Prickle3
Host Species Rabbit
Immunogen PRICKLE3 Fusion Protein Ag13632
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 317380, 4007, 54630
Target Species Rat, Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.