Learn More
Applied Biosystems™ PrepSEQ™ 1-2-3 Mycoplasma Nucleic Acid Extraction Kit
Product Code. 30000684 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
100 reactions
Unit Size:
100 reactions
Product Code. Quantity unitSize
30000684 100 reactions 100 reactions
1 options
Product Code. 30000684
Brand: Applied Biosystems™ 4443789
Edge
The Applied Biosystems PrepSEQ 1-2-3 Nucleic Acid Extraction Kit rapidly extracts Mycoplasma genomic DNA from 100 μL sample volumes of research cell lines, stem cell lines, and cell and tissue therapy samples.
The Applied Biosystems PrepSEQ 1-2-3 Nucleic Acid Extraction Kit rapidly extracts Mycoplasma genomic DNA from 100 μL sample volumes of research cell lines, stem cell lines, and cell and tissue therapy samples. In the PrepSEQ 1-2-3 kit protocol, a rapid procedure recovers DNA from total lysate of the test sample using magnetic-bead based technology. The PrepSEQ 1-2-3 kit is optimized for use with the MycoSEQ Mycoplasma Detection Kit.
Features of the PrepSEQ 1-2-3 Nucleic Acid Extraction Kit include:
  • High recovery of Mycoplasma genomic DNA from mammalian cell culture samples
  • Rapid easy-to-use protocol with results in under 2 hours
  • Compatible with the MycoSEQ Mycoplasma Detection assay
  • Protocol available for automated sample preparation
High DNA recovery sample preparation
Optimized for use with the MycoSEQ Mycoplasma Detection Kit, the PrepSEQ 1-2-3 Nucleic Acid Extraction Kit uses proprietary magnetic bead-based separation technology to extract Mycoplasma DNA from mammalian cell lines. The PrepSEQ 1-2-3 kit extracts Mycoplasma genomic DNA from research cell lines, stem cell lines, and cellular and tissue therapy samples with high efficiency.
The protocol is recommended for use with a starting sample volume of 100 μL with a maximum cell density of 106 cells. The PrepSEQ 1-2-3 kit provides an easy to use procedure consisting of 1) RNase treatment, 2) proteinase treatment, and 3) DNA extraction using the magnetic bead technology (figure below).
The kit isolates Mycoplasma genomic DNA with high sensitivity from total lysate of test sample. The PrepSEQ 1-2-3 kit and the MycoSEQ kit allow routine screening of high-value cell lines using an easy, rapid and sensitive protocol. The figure below demonstrates how a sample that was spiked with low levels of Mycoplasma bovoculi DNA was found negative for Mycoplasma contamination using a commercially prepared kit. When using the PrepSEQ 1-2-3 kit and the MycoSeq kit, the correct positive call for Mycoplasma contamination was made.
Specifications
Content And Storage Includes buffers, magnetic beads and enzymes for DNA extraction for Mycoplasma DNA extraction
Isolation Technology Magnetic Bead
Scale Mini
Sample Type Cell Culture Supernatants
Final Product Type Genomic DNA
High-throughput Compatibility High-throughput Compatible
No. of Reactions 100 Reactions
Product Line PrepSEQ
Quantity 100 reactions
Sample Volume 100 μL
Target Organism Class Mycoplasma spp.
Test Time 2 hr.