missing translation for 'onlineSavingsMsg'
Learn More

Pregnenolone, TRC

Product Code. 17999174
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17999174 50 mg 50mg
30136338 5 g 5g
30137327 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17999174 Supplier Toronto Research Chemicals Supplier No. TRCP71220050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145-13-1
Formula Weight 316.2402
InChI Formula InChI=1S/C21H32O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h4,15-19,23H,5-12H2,1-3H3/t15-,16-,17+,18-,19-,20-,21+/m0/s1
IUPAC Name 1-[(3S,8S,9S,10R,13S,14S,17S)-3-hydroxy-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]ethanone
Molecular Formula C21 H32 O2
Molecular Weight (g/mol) 316.48
Percent Purity >95%
SMILES CC(=O)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Synonym (3β)-3-Hydroxy-pregn-5-en-20-one, Pregn-5-en-20-one, 3β-hydroxy- (8CI), (3β)-3-Hydroxypregn-5-en-20-one, 17β-[1-Ketoethyl]-δ5-androsten-3β-ol, 3β-Hydroxy-5-en-20-one, 3β-Hydroxy-5-pregnene-20-one, 3β-Hydroxy-δ5-pregnen-20-one, 3β-Hydroxypregn-5-en-20-one, 5-Pregnenolone, Arthenolone, Bina-Skin, Enelone, NSC 1616, NSC 18158, Natolone, Pregn-5-en-3β-ol-20-one, Pregn-5-ene-3β-ol-20-one, Pregnelon, Pregnenolone, Pregnenolone (progesterone precursor), Pregnetan, Pregneton, Pregnolon, Prenolon, Regnosone, Skinostelon, δ5-Pregnen-3β-ol-20-one, δ5-Pregnene-3β-ol-20-one, δ5-Pregnenolone
Chemical Name or Material Pregnenolone
Grade HPLC
Show More Show Less

Specifications

Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.