Learn More
Honeywell™ Prefilter Pads
Product Code. 12682743
Change view
Click to view available options
Cartridge Type:
Bayonet Gas/Vapor Cartridges on N-Series Respirators
Click-Fit Cartridges on T-Series Respirators or on Freedom Series Respirators
Design Type:
Requires Adapter (N06575035)
Requires Retaining Cover (1001590 or 1001606)
For Use With (Application):
Respiratory Protection (P1, Dust)
Respiratory Protection (P2, Dust/Fume)
Respiratory Protection (P2/P3, Particulates)
Respiratory Protection (P2/P3, Particulates/Odor)
For Use With (Equipment):
Half-Mask (e.g. N5500™, N7700™) and Full-Face (e.g. N5400™) Air-Purifying Respirators
Half-Mask (e.g. Valuair™, Premier™, and MX/PF™) and Full-Face (e.g. Optifit Twin™) Air-Purifying Respirators
Unit Size:
1 pair
Each
Pack of 20
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cartridge Type Design Type For Use With (Application) For Use With (Equipment) unitSize
12682743 Click-Fit Cartridges on T-Series Respirators or on Freedom Series Respirators Requires Retaining Cover (1001590 or 1001606) Respiratory Protection (P2, Dust/Fume) Half-Mask (e.g. Valuair™, Premier™, and MX/PF™) and Full-Face (e.g. Optifit Twin™) Air-Purifying Respirators Each
11847171 Click-Fit Cartridges on T-Series Respirators or on Freedom Series Respirators Requires Retaining Cover (1001590 or 1001606) Respiratory Protection (P1, Dust) Half-Mask (e.g. Valuair™, Premier™, and MX/PF™) and Full-Face (e.g. Optifit Twin™) Air-Purifying Respirators Pack of 20
15806645 Bayonet Gas/Vapor Cartridges on N-Series Respirators Requires Adapter (N06575035) Respiratory Protection (P2/P3, Particulates/Odor) Half-Mask (e.g. N5500™, N7700™) and Full-Face (e.g. N5400™) Air-Purifying Respirators 1 pair
15896635 Bayonet Gas/Vapor Cartridges on N-Series Respirators Requires Adapter (N06575035) Respiratory Protection (P2/P3, Particulates) Half-Mask (e.g. N5500™, N7700™) and Full-Face (e.g. N5400™) Air-Purifying Respirators 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12682743 Supplier Honeywell™ Supplier No. 1002801
Only null left
Add to basket
Add to basket
Prefilter Pads are used in conjunction with a retaining clip or adapter on Honeywell™ Twin-Filter Air-Purifying Respirators
  • Product Type: Prefilter Pads
  • For Use With: Honeywell™ Twin-Filter Air-Purifying Respirators
TRUSTED_SUSTAINABILITY
Specifications
Cartridge Type Click-Fit Cartridges on T-Series Respirators or on Freedom Series Respirators
Certifications/Compliance PPE Cat. 3; ISO 9001/2000
Certifications (EN) EN 143
Design Type Requires Retaining Cover (1001590 or 1001606)
Filter Type P
Product Type Prefilter Pad
For Use With (Application) Respiratory Protection (P2, Dust/Fume)
For Use With (Equipment) Half-Mask (e.g. Valuair™, Premier™, and MX/PF™) and Full-Face (e.g. Optifit Twin™) Air-Purifying Respirators
Model TP2S
Quantity 1 Ea.
Weight (Metric) 0.01 kg