Learn More
Honeywell™ Prefilter Pads
Description
- Product Type: Prefilter Pads
- For Use With: Honeywell™ Twin-Filter Air-Purifying Respirators
Specifications
Specifications
| Cartridge Type | Click-Fit Cartridges on T-Series Respirators or on Freedom Series Respirators |
| Certifications/Compliance | PPE Cat. 3; ISO 9001/2000 |
| Certifications (EN) | EN 143 |
| Design Type | Requires Retaining Cover (1001590 or 1001606) |
| Filter Type | P |
| Product Type | Prefilter Pad |
| For Use With (Application) | Respiratory Protection (P2, Dust/Fume) |
| For Use With (Equipment) | Half-Mask (e.g. Valuair™, Premier™, and MX/PF™) and Full-Face (e.g. Optifit Twin™) Air-Purifying Respirators |
| Model | TP2S |
| Quantity | 1 Ea. |
| Show More |