missing translation for 'onlineSavingsMsg'
Learn More

Prednisone Glucuronide Sodium Salt, TRC

Product Code. 30412287
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.50mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412287 25 mg 25mg
30413865 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30412287 Supplier Toronto Research Chemicals Supplier No. TRCP70384025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Prednisone Glucuronide Sodium Salt
Analyte or Component Names Prednisone Glucuronide Sodium Salt
Shipping Condition Room Temperature
Molecular Formula C27H33NaO11
Synonym (2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-(2-((10R,13S,17R)-17-Hydroxy-10,13-dimethyl-3,11-dioxo-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthren-17-yl)-2-oxoethoxy)tetrahydro-2H-pyran-2-carboxylic Acid
InChI Formula InChI=1S/C27H34O11.Na/c1-25-7-5-13(28)9-12(25)3-4-14-15-6-8-27(36,26(15,2)10-16(29)18(14)25)17(30)11-37-24-21(33)19(31)20(32)22(38-24)23(34)35;/h5,7,9,14-15,18-22,24,31-33,36H,3-4,6,8,10-11H2,1-2H3,(H,34,35);/q;+1/p-1/t14?,15?,18?,19-,20-,21+,22-,24+,25-,26-,27-;/m0./s1
Solution Type Neat
SMILES O=C(CO[C@H]1[C@H](O)[C@@H](O)[C@H](O)[C@@H](C(O[Na])=O)O1)[C@]2(O)[C@@]3(C)CC(C4C(C3CC2)CCC5=CC(C=C[C@@]54C)=O)=O
Industry Type Forensics
Molecular Weight (g/mol) 556.53
Quantity 25 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.