missing translation for 'onlineSavingsMsg'
Learn More

Prednisolone 21-Methanesulfonate, TRC

Product Code. 30411753
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411753 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411753 Supplier Toronto Research Chemicals Supplier No. TRCP703765250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Prednisolone 21-Methanesulfonate
CAS 35410-28-7
Molecular Formula C22 H30 O7 S
Quantity 250 mg
Synonym 11β,17,21-Trihydroxypregna-1,4-diene-3,20-dione 21-Methanesulfonate, (11β)-11,17-Dihydroxy-21-[(methylsulfonyl)oxy]pregna-1,4-diene-3,20-dione
InChI Formula InChI=1S/C22H30O7S/c1-20-8-6-14(23)10-13(20)4-5-15-16-7-9-22(26,18(25)12-29-30(3,27)28)21(16,2)11-17(24)19(15)20/h6,8,10,15-17,19,24,26H,4-5,7,9,11-12H2,1-3H3/t15-,16-,17-,19+,20-,21-,22-/m0/s1
SMILES C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)COS(=O)(=O)C
IUPAC Name [2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] methanesulfonate
Molecular Weight (g/mol) 438.534
Formula Weight 438.171
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.