Learn More
Kern Sohn Precision Balance PLS
Click to view available options
Capacity:
120 g
420 g
720 g
4200 g
6200 g
8200 g
20 kg
Unit Size:
Each
Description
- Convenient recipe-weighing: with the recipe database, in which up to 99 recipes can be stored, each with up to 20 recipe ingredients with name and target value
- Rapid and efficient operation thanks to the graphics display
- Simple, clear user interface on the display in the following languages: German, English, French, Italian, Spanish, Portuguese
- Dosage aid: High stability mode and other filter settings can be selected
- Internal memory for complete recipes with name and target value of the recipe ingredients. User guidance through display
- Protective working cover included with delivery
Specifications
Specifications
| Capacity | 120 g |
| Readability | 0.001 g |
| Linearity | 0.003 g |
| Type | Precision Balance |
| Electrical Requirements | Universal Mains Adapter |