Learn More
Ambion™ Pre-miR™ miRNA Precursor
Product Code. p-7184675
Click to view available options
Quantity:
5 nmol
Unit Size:
Each
Product Code. 10782307
Brand: Ambion™ AM17100
Pre-miR™ miRNA Precursor
miRNA Precursor Molecules are small, chemically modified double-stranded RNA molecules designed to mimic endogenous mature miRNAs.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 5X
Content And Storage Pre-miR™ miRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C. It is guaranteed for 1 year when stored dried at –20°C.
Description Pre-miR™ miRNA Precursor Molecule, 5nmol, Liquid, Protect Against Physical Damage, Isolate From Incompatible Substances, Store in Original Container
Final Product Type Pre-miR™ miRNA Precursor Molecule
Packaging Type 1 Each
Product Line Ambion, Pre-miR
Product Type Pre-miR™ miRNA Precursor Molecule
Quantity 5 nmol
Sample Size 5nmol
Form Liquid