Learn More
Saint Gobain Life Sciences Rubber Stopper Without Holes
Click to view available options
Color:
Blue
Unit Size:
Pack of 5
Description
- Designed to keep container contents secure
- Ideal for securing glass, metal and plastic containers
- Easy to use
Specifications
Specifications
| Height (Metric) | 45mm |
| Quantity | 5/Pk. |
| Color | Blue |
| Diameter (Metric) Bottom | 56 mm |
| Diameter (Metric) Top | 65 mm |
| Material | Natural Rubber |
| Product Type | Rubber Stopper |