Learn More
Saint Gobain Life Sciences Pre-drilled Rubber Stopper Without Holes
Product Code. 10565814
Click to view available options
Color:
Blue
Unit Size:
Pack of 5
Product Code. 10565814
Brand: Saint Gobain Life Sciences 203083
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these stoppers for a variety of applications. Saint Gobain Life Sciences Rubber Stoppers Without Holes are ideal for chemical transfer.
  • Designed to keep container contents secure
  • Ideal for securing glass, metal and plastic containers
  • Easy to use
TRUSTED_SUSTAINABILITY
Specifications
Height (Metric) 60mm
Quantity 5/Pk.
Color Blue
Diameter (Metric) Bottom 71 mm
Diameter (Metric) Top 89 mm
Material Natural Rubber
Product Type Rubber Stopper