Learn More
Brady™ Pre-cut Vinyl Labels for BradyJet J4000 and J5000
Product Code. 17469192
Change view
Click to view available options
Height (Metric):
55.1 mm
74.9 mm
104.9 mm
177.8 mm
210 mm
Vertical Repeat (Metric):
92.1 mm
117.5 mm
162 mm
222.3 mm
266.7 mm
Width (Metric):
85.1 mm
110 mm
155 mm
254 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Vertical Repeat (Metric) Width (Metric) unitSize
17469192 210 mm 222.3 mm 155 mm Each
17499182 55.1 mm 92.1 mm 85.1 mm Each
17429192 74.9 mm 117.5 mm 110 mm Each
17439182 104.9 mm 162 mm 155 mm Each
17419192 177.8 mm 266.7 mm 254 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17469192 Supplier Brady™ Supplier No. 148664
Only null left
Add to basket
Add to basket
B-2569 is ideal for indoor facility labels, including 5S, safety, warning, pipe markers, equipment and visual workplace labels on stainless steel and smooth surfaces.
  • Specially-formulated topcoat enhances colors and label appearance
  • Tested to withstand exposure to liquids, alcohol and chemicals, as well as indoor fluorescent and UV lighting
  • Topcoat protects print from smudging and harsh conditions
  • Inkjet-receptive labels for use with the BradyJet J4000 or J5000 Color Label Printer
  • An ink cartridge is required to print this material
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Matte
Format Die-cut
Material Vinyl (Inkjet-receptive)
Printable Sides Single-side Printable
Print Technology Ink Jet
Product Type Label
Sterility Non-sterile
For Use With (Application) Electrical Labeling, Hazard Communication Identification, Warehouse Marking
For Use With (Equipment) BradyJet J4000 and J5000 Color Label Printers
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
No. per Pack 120 Labels/Roll
Quantity 120 Labels
Shape Rectangle
Temperature Range (English) -40°F to +140°F
Temperature Range (Metric) -40°C to +60°C
Thickness (English) 0.0053 in.
Thickness (Metric) 0.14 mm
Vertical Repeat (Metric) 222.3 mm
Dimensions (L x W) 210.82 x 154.94 mm
Length (English) 8.3 in.
Width (English) 6.1 in.
Width (Metric) 155 mm
Width (Metric) Printable Area 154.9 mm
Height (English) 8.3 in.
Height (Metric) 210 mm
Height (Metric) Printable Area 210 mm
Alternate Part Number J50-267-2595