Learn More
Brady™ Pre-cut Polyester Labels for BradyJet J4000 and J5000
Product Code. 17409202
Click to view available options
Height (Metric):
55.1 mm
74.9 mm
104.9 mm
177.8 mm
210 mm
Width (Metric):
85.1 mm
110 mm
155 mm
254 mm
Vertical Repeat (Metric):
92.08 mm
117.48 mm
161.93 mm
222.25 mm
266.70 mm
Unit Size:
Each
Product Code. 17409202
Brand: Brady™ 148668
Only null left
Add to basket
Add to basket
B-2569 is ideal for indoor facility labels, including 5S, safety, warning, pipe markers, equipment and visual workplace labels on stainless steel and smooth surfaces.
  • Specially-formulated topcoat enhances colors and label appearance
  • Tested to withstand exposure to liquids, alcohol and chemicals, as well as indoor fluorescent and UV lighting
  • Topcoat protects print from smudging and harsh conditions
  • Inkjet-receptive labels for use with the BradyJet J4000 or J5000 Color Label Printer
  • An ink cartridge is required to print this material
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Matte
Format Die-cut
Material Polyester (Inkjet-receptive)
Printable Sides Single-side Printable
Print Technology Ink Jet
Product Type Label
Sterility Non-sterile
For Use With (Application) Electrical Labeling, Hazard Communication Identification, Warehouse Marking
For Use With (Equipment) BradyJet J4000 and J5000 Color Label Printers
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
No. per Pack 120 Labels/Roll
Quantity 120 Labels
Shape Rectangle
Temperature Range (English) -40°F to +140°F
Temperature Range (Metric) -40°C to +60°C
Thickness (English) 0.0047 in.
Thickness (Metric) 0.11 mm
Vertical Repeat (Metric) 222.25 mm
Dimensions (LxW) 210.82 x 154.94 mm
Length (English) 8.3 in.
Width (English) 6.1 in.
Width (Metric) 155 mm
Width (Metric) Printable Area 154.9 mm
Height (English) 8.3 in.
Height (Metric) 210 mm
Height (Metric) Printable Area 210 mm
Alternate Part Number J50-267-2569