missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRDX3 Recombinant Rabbit Monoclonal Antibody (14E2)
GREENER_CHOICE

Product Code. 17978762
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17978762 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17978762 Supplier Invitrogen™ Supplier No. MA549994

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

PRDX3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow, ELISA

Peroxiredoxin (Prx) is a growing peroxidase family, whose mammalian members have been known to connect with cell proliferation, differentiation, and apoptosis. Many isoforms (about 50 proteins), collected in accordance to the amino acid sequence homology, containing active site cysteine residue, and the thiol-specific antioxidant activity, distribute throughout all the kingdoms. Among them, mammalian Prx consists of 6 different members grouped into typical 2-Cys, atypical 2-Cys Prx, and 1-Cys Prx. Except Prx VI belonging to 1-Cys Prx subgroup, the other five 2-Cys Prx isotypes have the thioredoxin-dependent peroxidase (TPx) activity utilizing thioredoxin, thioredoxin reductase, and NADPH as a reducing system. Mammalian Prxs are 20-30 kilodalton in molecular size and vary in subcellular localization: Prx I, II, and VI in cytosol, Prx III in mitochondria, Prx IV in ER and secretion, Prx V showing complicated distribution including peroxisome, mitochondria and cytosol.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRDX3
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 14E2
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PRDX3
Gene Accession No. P30048
Gene Alias antioxidant protein; antioxidant protein 1; anti-oxidant protein 1; Aop1; AOP-1; AW822249; D0Tohi1; Ef2l; HBC189; Mer5; mitochondrial thioredoxin dependent peroxide reductase; mitochondrial Trx dependent peroxide reductase; perioredoxin-3; peroxiredoxin 3; peroxiredoxin III; Peroxiredoxin-3; Prdx3; PRO1748; Protein MER5; Protein MER5 homolog; Prx; Prx 3; PRx III; Prx3; PRX-3; Prx-III; SP22; SP-22; TDXM; thioredoxin-dependent peroxide reductase, mitochondrial
Gene Symbols PRDX3
Host Species Rabbit
Immunogen A synthesized peptide derived from human PRDX3.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10935
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.