Learn More
Prazosin hydrochloride, MedChemExpress
Product Code. 16461396
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
Product Code. 16461396
Brand: Medchem Express HYB0193A
MedChemExpress Prazosin hydrochloride is a well-tolerated, CNS-active α1-adrenergic receptor antagonist for the research of high blood pressure and alcohol use disorders. Prazosin hydrochloride potently inhibits Norepinephrine (NE)-stimulated 45Ca efflux with an IC50 of 0.15 nM.Prazosin hydrochloride inhibits organic cation transporters OCT-1 and OCT-3 with IC50s of 1.8, and 13 μM, respectively.
Prazosin hydrochloride is a well-tolerated, CNS-active α1-adrenergic receptor antagonist for the research of high blood pressure and alcohol use disorders. Prazosin hydrochloride potently inhibits Norepinephrine (NE)-stimulated 45Ca efflux with an IC50 of 0.15 nM.Prazosin hydrochloride inhibits organic cation transporters OCT-1 and OCT-3 with IC50s of 1.8, and 13 μM, respectively.
Specifications
Chemical Name or Material Prazosin hydrochloride
CAS 19237-84-4
Color White
Purity Grade Notes Research
Molecular Formula C19H22ClN5O4
Quantity 200 mg
Solubility Information DMSO : 14.29 mg/mL (34.04 mM; Need ultrasonic) ∣H2O : 0.59 mg/mL (1.41 mM; Need ultrasonic)
SMILES O=C(N1CCN(C2=NC(N)=C3C=C(OC)C(OC)=CC3=N2)CC1)C4=CC=CO4.Cl
Molecular Weight (g/mol) 419.86
Formula Weight 419.86
Percent Purity 99.93%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation