missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPP2R3B Polyclonal Antibody
GREENER_CHOICE

Product Code. 16589911
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16589911 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16589911 Supplier Invitrogen™ Supplier No. PA597082

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: FSGAVTRGRK VQKEGKISYA DFVWFLISEE DKKTPTSIEY WFRCMDLDGD GALSMFELEY FYEEQCRRLD SMAIEALPFQ DCLCQMLDLV KPRTEGKITL QDLKRCKLAN VFFDTFFNIE KYLDHEQKEQ ISLLRDGDSG GPELSDWEKY AAEEYDILVA EETAGEPWED GFEAELSPVE QKLSALRSPL AQRPFFEAPS PLGAVDLYEY ACGDEDLEPL; Positive Samples: HeLa, SW620, BT-474, SKOV3, mouse skeletal muscle; Cellular Location: Nucleus.

Protein phosphatase 2 is one of the four major Ser/Thr phosphatases and is implicated in the negative control of cell growth and division. Protein phosphatase 2 holoenzymes are heterotrimeric proteins composed of a structural subunit A, a catalytic subunit C, and a regulatory subunit B. The regulatory subunit is encoded by a diverse set of genes that have been grouped into the B/PR55, B'/PR61, and B' '/PR72 families. These different regulatory subunits confer distinct enzymatic specificities and intracellular localizations to the holozenzyme. The product of this gene belongs to the B' ' family. The B' ' family has been further divided into subfamilies. The product of this gene belongs to the beta subfamily of regulatory subunit B' '. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R3B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PPP2R3B
Gene Accession No. Q9Y5P8
Gene Alias NYREN8; NY-REN-8; NY-REN-8 antigen; PP2A subunit B isoform PR48; PP2A, subunit B, PR48 isoform; PPP2R3B; PPP2R3L; PPP2R3LY; PR48; protein phosphatase 2 (formerly 2A), regulatory subunit B' ', beta; protein phosphatase 2 regulatory subunit B' ', beta; protein phosphatase 2 regulatory subunit B' 'beta; protein phosphatase 2, regulatory subunit B' ', beta; Protein phosphatase 2A 48 kDa regulatory subunit; serine/threonine protein phosphatase 2A, 48kDa regulatory subunit B; serine/threonine-protein phosphatase 2A regulatory subunit B' ' subunit beta
Gene Symbols PPP2R3B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 356-575 of human PPP2R3B (NP_0373712).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28227
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.