missing translation for 'onlineSavingsMsg'
Learn More

PPP1R13L Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16866405 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16866405 150 μL 150µL
16876405 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16866405 Supplier Proteintech Supplier No. 511411AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes paired box gene 6, one of many human homologs of the Drosophila melanogaster gene prd. In addition to the hallmark feature of this gene family, a conserved paired box domain, the encoded protein also contains a homeo box domain. Both domains are known to bind DNA, and function as regulators of gene transcription. This gene is expressed in the developing nervous system, and in developing eyes. Mutations in this gene are known to cause ocular disorders such as aniridia and Peter′s anomaly. Alternatively spliced transcript variants encoding either the same or different isoform have been found for this gene.

Specifications

Antigen PPP1R13L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PPP1R13L
Gene Accession No. Q5I1X5, Q8WUF5
Gene Alias IASPP, Inhibitor of ASPP protein, NKIP1, PPP1R13B like protein, PPP1R13BL, PPP1R13L, Protein iASPP, RAI, RelA associated inhibitor
Gene Symbols Ppp1r13l
Host Species Rabbit
Immunogen PPP1R13L Fusion Protein Ag0635
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10848, 333654
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.