missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPEF2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
461.00 EUR
Specifications
| Antigen | PPEF2 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
PPEF2 Polyclonal specifically detects PPEF2 in Human samples. It is validated for Western Blot.Specifications
| PPEF2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Vision | |
| PBS buffer, 2% sucrose | |
| 5470 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| protein phosphatase 7, catalytic subunit, beta isozyme, protein phosphatase with EF hands 2, protein phosphatase, EF hand calcium-binding domain 2, protein phosphatase, EF-hand calcium binding domain 2, serine/threonine-protein phosphatase with EF-hands 2 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Human PPEF2 (NP_006230). Peptide sequence LVTGEKEEPSRSASEADSEAGELRKPTQEEWRQVVDILWSDPMAQEGCKA | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title