Learn More
Hirschmann™ Polypropylene Funnels
Product Code. 10286581
Change view
Click to view available options
Diameter (Metric)Top:
40mm
75mm
100mm
120mm
150mm
Unit Size:
Pack of 12
Pack of 24
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric)Top unitSize
10286581 120mm Pack of 12
10358231 100mm Pack of 12
10184341 150mm Pack of 12
10092751 40mm Pack of 24
10308671 75mm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10286581 Supplier Hirschmann™ Supplier No. 7750107
Only null left
Add to basket
Add to basket
For use with various laboratory applications
  • Transparent
Specifications
Diameter (Metric)Top 120mm
Material Polypropylene
Height (Metric) 180 mm
Length (Metric) Stem 90mm
Quantity 12/Pk.