Learn More
SymLine™ Power Supply for SignalBox2
Product Code. 17455053 Shop All SCAT Europe Products
Click to view available options
Color:
Black
Unit Size:
Each
Product Code. 17455053
Brand: SymLine™ 108326
Only null left
Add to basket
Add to basket
Power supply for use with the built-in SignalBox2.
  • EU power supply unit for the power supply of the SCAT™ Level Control (220/230 V) to be reordered
  • Length of the cable: 1900 mm
Specifications
Color Black
For Use With (Equipment) Built-in SignalBox2
Product Type Power Supply