Learn More
Karl Hecht™ Powder Funnel, Polypropylene
Click to view available options
Diameter (Metric)Top:
100mm
120mm
150mm
Unit Size:
Each
Description
- Polypropylene construction.
- Transparent.
- Facilitates pouring operations.
- Reduces the risk of spilling.
Specifications
Specifications
| Diameter (Metric)Top | 150mm |
| Material | Polypropylene |
| Product Type | Funnel |