Learn More
Borer Chemie™ deconex 24 FORTE
Product Code. 10084470
Click to view available options
Volume (Metric):
1 x 10 kg
Unit Size:
10kg
Product Code. 10084470
Brand: Borer Chemie™ 501900.00S10P
Powder cleaner for automated cleaning of laboratory glassware and utensils. For the highest demands and stubborn residues, deconex 24 FORTE is suitable. An acidic rinse is recommended. Contains active chlorine as a cleaning booster.
  • Powder cleaner for automated cleaning
  • With active chlorine
  • Typical application concentration: 3-5 g/L
  • Can be rinsed off without leaving residues
  • For consistent and flawless results
  • Wide cleaning spectrum
  • Without EDTA, NTA and surfactants
  • Acid rinse with deconex cleaner recommended
  • Rechnical support by the manufacturer
Specifications
Form Powder
pH ∼12.3 (1% Solution)
Composition Alkalis, complexing agents, corrosion inhibitor, organic chlorine donor
Concentration Concentrate
Container Type Bucket
Content And Storage 5°C to 25°C
Quantity 1 x 10 kg
Volume (Metric) 1 x 10 kg
Weight (English) 22 lb.
Weight (Metric) 10 kg
compliance-icons
Product Identifier
  • deconex 24 LIQ
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : potassium hydroxide,dipotassium trioxosilicate, sodium hypochlorite