Learn More
Miele Professional™ Potential-Free Contacts PLW8636 APLW 882
Shop All Miele Products
Click to view available options
Product Type:
Pump
Unit Size:
1 sheet
Description
- 10 potential-free contacts (4 inputs and 6 outputs)
- For external pumps, valves, fans and ring line systems, for example
- Installed by Customer Service
- Activating external signal receivers for PLW 8636
Specifications
Specifications
| Product Type | Pump |
| For Use With (Equipment) | Washer-Disinfectors |
| Quantity | 1 Ea. |