Learn More
Memmert™ Potential-Free Contact (24 V / 2 A) with Socket to NAMUR NE 28 for Combined Error Message for IN160
Product Code. 17527475 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IN160 Incubator
Unit Size:
Each
Product Code. 17527475
Brand: Memmert™ IN160H6
Only null left
Add to basket
Add to basket
Potential-free contact for combination error message (e.g. supply failure, sensor fault, fuse).
  • Accessory
  • For combination error message
Specifications
For Use With (Equipment) IN160 Incubator
Type Potential-free Contact
Includes Socket to NAMUR NE 28
Voltage 24 V
Amperage 2 A