Learn More
Potassium Thiocyanate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber glass bottle
HDPE plastic bottle
Unit Size:
1 L
2.5 L
Specifications
Specifications
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 1 L |
| Packaging | Amber glass bottle |