missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium permanganate concentrate, For 1L standard solution, 1/500 M KMnO4 (0.01N), Honeywell Fluka™
Shop All Honeywell Fluka ProductsDescription
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Physical Form | Concentrate |
| Quantity | 6 x 1 Ea. |
| Linear Formula | KMnO4 |
| UN Number | NONH for all modes of transport |
| SMILES | [K+].[O-][Mn](=O)(=O)=O |
| IUPAC Name | potassium manganesoylolate |
| Molecular Weight (g/mol) | 158.03 |
| Formula Weight | 158.03 |
| Packaging | Ampoule |
| Chemical Name or Material | Potassium permanganate concentrate, For 1L standard solution, 1/500 M KMnO4 (0.01N) |
Safety and Handling
Product Identifier
- Potassium permanganate concentrate
- LONG-TERM AQUATIC HAZARD Chronic 3
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contain: potassium permanganate
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?