missing translation for 'onlineSavingsMsg'
Learn More

Potassium Permanganate, 0.02M/l Solution, Scharlab

Product Code. 11314284
Click to view available options
Quantity:
1 L
Unit Size:
1 L
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

Product Code. 11314284

Brand: Scharlab PO03361000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

Used in analytical chemistry as a titrant in volumetric analysis.

This volumetric solution was checked by means of potentiometric methods using Scharlau's disodium oxalate volumetric standard. Scharlau's volumetric standards are directly traceable to the Standard Reference Materials from NIST (National Institute of Standards and Technology, USA).

  • Molecular Formula: KMnO4
  • Molar Mass: 158.04g/mol
  • CAS: 7722-64-7
  • EIN ECS No.: 231-760-3
  • Factor: 0.999–1.001

Specifikationer

CAS 7722-64-7
Physical Form Solution
Quantity 1 L
Molecular Formula KMnO4
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
SMILES [O-][Mn](=O)(=O)=O.[K+]
IUPAC Name potassium;permanganate
Molecular Weight (g/mol) 158.032
PubChem CID 516875
Formula Weight 158.04g/mol
Density 1.01g/cm3
Visa mer Visa mindre
Produktbeteckning
  • Potassium permanganate, solution 0,02 mol/l (1/N)
Riskkategorier
  • LONG-TERM AQUATIC HAZARD Chronic 3
Faroangivelser
  • H412-Harmful to aquatic life with long lasting effects.
Skyddsangivelse
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Kompletterande information
  • MIXTURE LIST-Contains : Potassium permanganate
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.