Learn More
Potassium 4-(benzyloxycarbonylamino)phenyltrifluoroborate, 96%, Thermo Scientific™
Product Code. 15459955
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15459955 1 g 1g
15469955 5 g 5g
2 options
Product Code. 15459955
Brand: Thermo Scientific Alfa Aesar H52621.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 850623-45-9
Molecular Formula C14H12BF3KNO2
Molecular Weight (g/mol) 333.159
MDL Number MFCD04115743
InChI Key ADBHDDIXRRMUNK-UHFFFAOYSA-N
Synonym potassium 4-cbz-aminophenyl trifluoroborate, potassium 4-benzyloxycarbonylamino phenyltrifluoroborate, potassium 4-benzyloxy carbonyl amino phenyl trifluoroborate, potassium 4-benzyloxy carbonyl amino phenyl trifluoroboranuide, potassium 4-carboxybenzoylaminophenyl trifluoroborate, potassium 4-benzyloxy carbonyl amino phenyl trifluoro borate 1-
PubChem CID 24851538
IUPAC Name potassium;trifluoro-[4-(phenylmethoxycarbonylamino)phenyl]boranuide
SMILES [B-](C1=CC=C(C=C1)NC(=O)OCC2=CC=CC=C2)(F)(F)F.[K+]
Specifications
Melting Point 250°C to 266°C
Quantity 1 g
Formula Weight 333.16
Percent Purity 96%
Chemical Name or Material Potassium 4-(benzyloxycarbonylamino)phenyltrifluoroborate