missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ViewRNA™ Tissue Positive Control Kit (1-plex) (4 rat kidney slides + probe)
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Description
The ViewRNA Tissue Positive Control Kit (1-Plex) is designed for use with the ViewRNA Tissue Assay Core Kit, 1-plex (Cat. No. 19931). This positive control kit includes four each of 5-μm thick rat kidney FFPE sections attached to positively charged slides as well as TYPE 1 Rat Ubc Probe Set.
Specifications
Specifications
| Content And Storage | • 5 μm thick rat kidney FFPE sections attached to positively charged slides, 4 slides • TYPE 1 Rat Ubc Probe Set |
| Format | Kit |
| For Use With (Application) | Tissue Analysis |
| For Use With (Equipment) | ViewRNA Tissue Assay Core Kit, 1-plex (Cat. No. 19931) |
| Product Type | RNA In Situ Hybridization Tissue Positive Control Kit |
| Quantity | 1 Kit |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction