Learn More
Haldenwanger™ Porcelain Desiccator Plates
Click to view available options
Diameter (Metric) Outer:
90 mm
140 mm
190 mm
235 mm
280 mm
Unit Size:
Each
Description
- Available in a variety of sizes
- Complies with DIN VDE 0335, type C 100
Specifications
Specifications
| Diameter (Metric) Outer | 190 mm |
| No. per Pack | 1 Each |
| Product Type | Desiccator Plate |
| Material | Porcelain |
| For Use With (Application) | Above desiccators. |