Learn More
Haldenwanger™ Porcelain Buchner Funnel
Product Code. p-8000239
Click to view available options
Capacity (Metric):
50 mL
75 mL
135 mL
290 mL
580 mL
795 mL
Diameter (Metric) Plate:
48mm
59mm
74mm
90mm
114mm
129mm
Unit Size:
Each
Product Code. 10433281
Brand: Haldenwanger™ L 127C/0A
Only null left
Add to basket
Add to basket
Designed with glazed porcelain for excellent thermal and chemical resistance. Haldenwanger™ Porcelain Buchner Funnels are an excellent choice for quick filtration.
  • Vertical sides
  • Available in a variety of sizes to fit many filtration needs
  • Perforated fixed filter plate
Specifications
Capacity (Metric) 50 mL
Material Porcelain
Diameter (Metric) Plate 48mm
Quantity 1Pack
Product Type Buchner Funnel