missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ POP™ Conformational Analysis Polymer (CAP)
Shop All Applied Biosystems Products
Click to view available options
Quantity:
25 mL
Unit Size:
Each
Description
This POP™ polymer is a Conformational Analysis Polymer for non-denaturing polynucleotide fragment analysis applications on Genetic Analyzers.
Order Info
Each
Specifications
Specifications
| Content And Storage | Refrigerate at 2°C to 8°C. |
| For Use With (Equipment) | 310 Genetic Analyzer, 3100 Genetic Analyzer, 3130xl Genetic Analyzer, 3730xl DNA Analyzer, 3130 Genetic Analyzer, 3730 DNA Analyzer |
| For Use With (Application) | Sequencing, Fragment Analysis (Non-Denaturing) |
| On Instrument Life Usage | Stability on instrument 7 days |
| Product Type | Sequencing Reagent |
| Quantity | 25 mL |
| Shipping Condition | Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction