Learn More
Thermo Scientific™ Ponceau S Staining Solution
Product Code. 30088667
Change view
Click to view available options
Quantity:
500 mL
1000 mL
3 x 1000 mL
Unit Size:
1000mL
500mL
Pack of 3
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30088667 3 x 1000 mL Pack of 3
17435103 500 mL 500mL
17641536 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30088667 Supplier Thermo Scientific™ Supplier No. A40000278X3
Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot.
Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot. This stain is compatible with nitrocellulose and PVDF membranes. Ponceau S staining is reversible and can be removed with a short incubation in 0.1% NaOH.
Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot. This stain is compatible with nitrocellulose and PVDF membranes. Ponceau S staining is reversible and can be removed with a short incubation in 0.1% NaOH.
Features include:
• A nondestructive, reversible method to stain and detect proteins on nitrocellulose and PVDF membranes
• Minimal nonspecific interactions with the membrane surface; provides a reliable method for visualization of protein transfer to a membrane
• Simple and quick staining protocol that results in reddish-pink bands that are easily photographed
• Reversed easily in less than five minutes
• Subsequent western blot detection unaffected as the stain does not alter the membrane-bound protein
TRUSTED_SUSTAINABILITY
Specifications
Detection Method Colorimetric
Formulation 0.1% (w/v) in 5% acetic acid
For Use With (Application) Western Blotting
Quantity 3 x 1000 mL
Product Type Ponceau S Staining Solution