Learn More
Kinematica™ Polytron™ Homogenizer Stands: Safety Rings
Product Code. 10233152 Shop All Kinematica Products
Click to view available options
For Use With (Equipment):
ST-F20/600, PT-ST 21
ST-P10/600 Stand
Unit Size:
Each
Product Code. 10233152
Brand: Kinematica™ 11045031
Prevents homogenizer from moving out of position.
The safety ring prevents uncontrolled lowering of the drive unit. Can also be used to fixing the drive unit height.
Specifications
Product Type Safety Positioning Ring
Material Stainless Steel
For Use With (Equipment) ST-F20/600, PT-ST 21