Learn More
Honeywell™ POLYTRIL™ Gloves
Description
- White polyamide knitted glove
- Gray oilproof nitrile coating on palm and fingertips
- Elastic knitted wrist
- The knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
- The glove offers maximum comfort and added sturdiness
- The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
- The ventilated back improves the glove's breathability
- The knitted and elasticated wrist gives the glove good support
- The nitrile coating at the palm and fingertips provides outstanding abrasion resistance and impermeability to oils
- Its gray coating makes the glove more soilproof, thus increasing its last
Specifications
Specifications
| Certifications/Compliance | EN 420-03 EEC Basic standards, EN 388-16 Mechanical hazards |
| Description | 13 gauge |
| Coating Material | Polyamide, Nitrile |
| Color | Gray, White |
| Size | 9 |
| Thickness (Metric) | 0.25 mm |
| Length (Metric) | 24.5 cm |