Learn More
Buerkle™ Polystyrene Volumetric Spoons
Shop All Bürkle Products
Click to view available options
Capacity (Metric):
0.5 mL
1 mL
Unit Size:
Pack of 100
Description
- Polystyrene, blue
- Round shape, easy emptying
Specifications
Specifications
| Capacity (Metric) | 1 mL |
| Color | Blue |
| Product Type | Volumetric Spoon |
| Material | Polystyrene |