Learn More
Fisherbrand™ Polystyrene Semi Micro Cuvettes for UV and Visible Wavelengths
Product Code. 11827922 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
1.6 mL
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11827922 1.6 mL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11827922
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these cuvettes for more constant sample temperatures during photometric measurements. Fisherbrand™ Polystyrene Semi Micro Cuvettes for UV and Visible Wavelengths feature excellent optical transmission range and small wall thickness for increased heat transfer.
  • Very low variation of extinction values
  • Excellent optical transmission range
  • Cavity-sorted production
Specifications
Type Semi-micro Cuvette
Disposable Reusable
Capacity (Metric) 1.6 mL
Material Plastic
Quantity 100/Pk.
Wavelength Range 220 to 900 nm
Height (Metric) 45 mm
Width (Metric) 12.5 mm