Learn More
Gosselin™ Polystyrene Round Petri Plate with One Vent
Click to view available options
Diameter (Metric):
90 mm
Unit Size:
Pack of 700
Description
- Used in variety of applications like clinical, environment, and microbiology
Specifications
Specifications
| Certifications/Compliance | EC N 1935/2004, ISO 11137 : Aseptic |
| Color | Natural |
| Material | Polystyrene |
| Shape | Round |
| Sterility | Aseptically Manufactured |
| Well Count | 1 |
| Quantity | 28/Bag |
| Vent | 1 |
| Height (Metric) | 16.2 mm |
| Diameter (Metric) | 90 mm |