Learn More
Gosselin™ Polystyrene Round Petri Plate with One Vent
Product Code. 15453684
Click to view available options
Diameter (Metric):
90 mm
Unit Size:
Pack of 700
Product Code. 15453684
Brand: Gosselin™ SH91101
Only null left
Add to basket
Add to basket
Used in variety of applications like clinical, environment, industries and microbiology.
  • Used in variety of applications like clinical, environment, and microbiology
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EC N 1935/2004, ISO 11137 : Aseptic
Color Natural
Material Polystyrene
Shape Round
Sterility Aseptically Manufactured
Well Count 1
Quantity 28/Bag
Vent 1
Height (Metric) 16.2 mm
Diameter (Metric) 90 mm