Description
Compatible with automated systems. U-shaped inner well contour for easy identification
- U-shaped inner well contour for easy identification
- Minimal bending of skirts
- Continuous skirt for compatibility with automated systems
- Stackable
- 8 holes in the skirt aid plate positioning and removal from the thermal cycler block e.g., with grippers
- Within SBS footprint recommendations (127.76 ±0.25mm x 85.47 ±0.25mm)
- Suitable for all common 384 well thermocyclers
- 45μL max well volume
Specifications
Specifications
| Barcode | Includes Barcode |
| Color | Blue |
| Material | Polypropylene |
| No. of Wells | 384 |
| Total Volume (Metric) Well | 45 μL |
| Well Shape | Round Bottom (U-well) |
| No. per Pack | 25 |
| Plate Compatibility | Automated systems |
| Quantity | 25 Pack |