Learn More
Kartell™ Polypropylene Stirring Paddles
Product Code. 10566961
Change view
Click to view available options
Size:
39 mm W x 15 mm H
57 mm W
65 mm W x 78 mm H
98.5 mm W
Unit Size:
Each
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
10566961 57 mm W Pack of 5
11308623 39 mm W x 15 mm H Each
11368623 65 mm W x 78 mm H Each
10294471 98.5 mm W Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10566961 Supplier Kartell™ Supplier No. 0043100
Only null left
Add to basket
Add to basket
Constructed of polypropylene and steel for strength, durability and chemical resistance. Kartel™ Polypropylene Stirring Paddles provide a sturdy variety of paddles to fit any stirring need.
  • Steel rod covered with polypropylene provides a strong connection to the stirrer
  • Steel rods: 8 dia. x 350mmL fits standard overhead stirrer
  • Polypropylene blades are premanently fixed to the rods for stability
    • Fixed propeller for vortex stirring
    • U shape for slower but more vigorous agitation
    • Two sizes of swivel impellers enable the blades to be introduced through a small aperture then fully extend for stirring
Specifications
Description Collapsible paddle, fully extends during stirring
Size 57 mm W
Type Stirring Paddle
Width (Metric) 57 mm
Material Polypropylene Paddle, Steel Rod Covered with Polypropylene