Learn More
BD Polypropylene Sharps Disposal Containers
Product Code. 15212139
Change view
Click to view available options
Capacity (Metric):
2.4 L
Dimensions (L x W x H):
182 x 181 x 260 mm
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Dimensions (L x W x H) unitSize
15212139 2.4 L 182 x 181 x 260 mm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15212139 Supplier BD Supplier No. 305625
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Offers a user-friendly option for the safe and proper disposal of sharp objects. BD™ Polypropylene Sharps Disposal Containers are lidded bins available in a variety of sizes. These safety products can be placed on counters, transported to various sites or wall mounted.
Proper disposal of sharp and pointed objects is the first step to a safer working environment. If they are not properly disposed of, these sharp and sharp objects may injure medical personnel or other persons. On site disposal helps to reduce needle stick injuries and blood contact risk.
  • Easy to use for safe operation and available in various sizes
  • Easy to attach container lid with visible temporary or permanent closure
  • Made of autoclavable polypropylene with minimum wall thickness of 2mm
  • Supplied with handle for easier transportation or brackets for wall mounting
  • Complies with the standards BS 7320, NF NX-500, UN-ADR 2392
  • Developed in accordance with the European Waste List Class 18.01.03
Specifications
Capacity (Metric) 2.4 L
Certifications/Compliance CE BS 7320, NF NX-500, UN-ADR 2392
Closure Type Snap-on
Closure Color White
Color Yellow
Dimensions (L x W x H) 182 x 181 x 260 mm
Disposable Single-use
Quantity 25 Pk.
Autoclavable Autoclavable
Includes Mounting brackets
For Use With (Application) For Containment of Sharps Waste