Learn More
Kautex™ Polypropylene Screw Closure for Series 310 Safegrip Bottles
Click to view available options
Thread Size (Metric):
00 mm
Unit Size:
Pack of 100
Description
- Manufactured from polypropylene
- Blue in color
- With PTFE insert and tamper-evident ring
- Applications include storage, packaging and transport
- Acc. to DIN 32
- Fits bottle capacities from 50 to 500 mL
Specifications
Specifications
| Color | Blue |
| For Use With (Application) | Packaging, Transport, Storage |
| Material | Polypropylene |
| No. per Pack | 100/Pk. |
| Thread Size (Metric) | 00 mm |
| Liner Material | PTFE |
| Neck Finish | 32 mm |
| Certifications/Compliance | DIN 32 |
| Product Type | Screw Cap |
| For Use With (Equipment) | Series 310 Safegrip Bottles |