Learn More
JG Finneran Associates™ Polypropylene Screw Caps
Product Code. 12452900
Click to view available options
Color:
Black
Red
Closure Size:
10-425
13-425
8-425
Type:
Large Open Hole Top Screw Cap
Open Top Screw Cap
Screw Cap
Unlined Solid Top Screw Cap
Unit Size:
Pack of 1000
Product Code. 12452900
Brand: JG Finneran Associates™ 531008R
Only null left
Specifications
Color Red
Material Polypropylene
Septum Molded
Style Large Open Hole Top Screw
Quantity 1000 Pk.
Type Screw Cap
Closure Size 8-425
For Use With (Equipment) Shimadzu with Flange, Dram Vials