Learn More
Menshen™ Polypropylene Screw Caps Wide Neck, Blue
Click to view available options
Certifications/Compliance:
DIN 54
Unit Size:
Each
Description
- Blue
- For 500-1000ml bottle
- PP
Specifications
Specifications
| Color | Blue |
| Material | Polypropylene |
| No. per Pack | 1 Each |
| Quantity | 1 Each |
| Screw Color | Blue |
| Screw Material | Polypropylene |
| Closure Color | Blue |
| Certifications/Compliance | DIN 54 |
| Product Type | Screw Cap |
| Closure Material | Polypropylene |
| Show More |