Learn More
Gilac™ White Polypropylene Scoop
Product Code. 13471989
Click to view available options
Capacity (Metric):
500 mL
750 mL
1000 mL
2 L
Unit Size:
Each
Product Code. 13471989
Brand: Gilac™ G601121
Only null left
Add to basket
Add to basket
White Polypropylene Scoop is a flour scoop, but is also convenient for all types of ingredients.
  • Constructed of polypropylene
  • For all types of ingredients
  • BPA Free
Specifications
Capacity (English) 16.9 oz.
Capacity (Metric) 500 mL
Color White
Product Type Scoop
Length (English) 11.2 in.
Size 29 cm
Material Polypropylene
Length (Metric) 285 mm