Learn More
Azpack™ Polypropylene Round Buckets with Plastic Handle
Product Code. 13199310
Click to view available options
Capacity (Metric):
1.18 L
15 L
Diameter (Metric) Outer:
105 to 132 mm
318 mm
Height (Metric):
131 mm
292.5 mm
Unit Size:
Each
Pack of 5
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Product Code. 13199310
Brand: Azpack™ 13199310
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Store, lift, and transport food items and other contents with less effort. Azpack™ Polypropylene Round Buckets with Plastic Handle are also suitable for collecting, carrying and storing biological samples.
  • Constructed of white food grade polypropylene
  • UN approved
  • Supplied with handles and lids
  • Suitable for packaging food
Specifications
Product Type Bucket
Capacity (Metric) 1.18 L
Material Polypropylene
Color White
Diameter (Metric) Outer 105 to 132 mm
Handle Material Plastic
Height (Metric) 131 mm
Includes Handle and Lid
Quantity 5 Pk.