Learn More
Cole-Parmer™ Colder™ Quick-Disconnect Fittings
Product Code. 11769619
Click to view available options
For Use With:
0.625 NPT(M) Thread x 0.625 in. tubing I.D.
3/8 in. tubing I.D.
Unit Size:
Each
Product Code. 11769619
Brand: Cole-Parmer™ 3160331
Eliminate virtually any loss of product with this leak-free fitting. Colder™ Quick-Disconnect Fittings available in a variety of materials and configurations and feature excellent chemical resistance.
For a complete fitting, order one body and one insert. All bodies will mate with inserts to make a fitting that will work for multiple applications.
  • Less than 0.10mL spillage
  • One-hand operation
Specifications
Material Polypropylene
For Use With 3/8 in. tubing I.D.
Packaging Quantity 1 Each
Includes Less than 0.10mL spillage
Product Type Quick Disconnect Fitting